4-Brombenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brombenzolsulfonsäureamid

16374-5G
SIAL16374-5GEA 70.3 EUR
SIAL16374-5G
4-Brombenzolsulfonamid, Sigma-Aldrich®
4-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00051977
CAS-Nummer: 701-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR