3-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

164135-25G
SIAL164135-25GEA 41.1 EUR
SIAL164135-25G
3-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®
3-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
MDL: MFCD00005430
CAS-Nummer: 5834-16-2
EINECS: 227-418-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR