7-Methoxy-2-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-7-methoxynaphthalen-2(1H)-one

164186-5G
SIAL164186-5GEA 92.8 EUR
SIAL164186-5G
7-Methoxy-2-tetralon, Sigma-Aldrich®
7-Methoxy-2-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD00001730
CAS-Nummer: 4133-34-0
EINECS: 223-954-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR