Trityliumhexafluorphosphat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Triphenylcarboniumhexafluorophosphat
M-Clarity™ Qualitätsstufe = MQ200
164569-10G
SIAL164569-10GEA
192
EUR
SIAL164569-10G
Trityliumhexafluorphosphat, Sigma-Aldrich®
Trityliumhexafluorphosphat
Formel:
C₁₉H₁₅F₆P Molecular Weight: 388,29 g/mol Schmelzpunkt: 166…167 °C |
MDL:
MFCD00013121 CAS-Nummer: 437-17-2 EINECS: 207-112-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...