Tropyliumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ200

164623-10G
SIAL164623-10GEA 769 EUR
SIAL164623-10G
Tropyliumtetrafluoroborat, Sigma-Aldrich®
Tropyliumtetrafluoroborat
Formel: C₇H₇BF₄
Molecular Weight: 177,94 g/mol
MDL: MFCD00013402
CAS-Nummer: 27081-10-3
EINECS: 248-214-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR