Methyl-5-chlorsalicylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 5-chloro-2-hydroxybenzoate

M-Clarity™ Qualitätsstufe = MQ100

165018-100G
SIAL165018-100GEA 208 EUR
SIAL165018-100G
Methyl-5-chlorsalicylat, Sigma-Aldrich®
Methyl-5-chlorsalicylat
Formel: C₈H₇ClO₃
Molecular Weight: 186,60 g/mol
MDL: MFCD00016461
CAS-Nummer: 4068-78-4
EINECS: 223-778-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR