DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane) ≥85% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: DPDPB

M-Clarity™ Qualitätsstufe = MQ100

16646-10MG-F
SIAL16646-10MG-FEA 127 EUR
SIAL16646-10MG-F
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane) ≥85% (durch HPLC), Sigma-Aldrich®
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)

1,4-Bis[3-(2-pyridyldithio)propionamido]butane can be used to study protein structural analysis, protein modification, proteomics and homobifunctional cross-linking reagents.


  • Reagent for protein-protein cross-linking via cysteine crosslinking


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₀H₂₆N₄O₂S₄
Molecular Weight: 482,71 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153604
CAS-Nummer: 141647-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR