Methyl-2,5-dichlorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,5-dichlorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

166731-10G
SIAL166731-10GEA 73.8 EUR
SIAL166731-10G
Methyl-2,5-dichlorbenzoat, Sigma-Aldrich®
Methyl-2,5-dichlorbenzoat
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00000606
CAS-Nummer: 2905-69-3
EINECS: 220-815-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR