2,2'-Dithiodianilin (Thulium-Ionophor I), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2-aminophenyl)disulfid

M-Clarity™ Qualitätsstufe = MQ100

166766-10G
SIAL166766-10GEA 123 EUR
SIAL166766-10G
2,2'-Dithiodianilin (Thulium-Ionophor I), Sigma-Aldrich®
2,2'-Dithiodianilin (Thulium-Ionophor I)
Formel: C₁₂H₁₂N₂S₂
MDL: MFCD00007703
CAS-Nummer: 1141-88-4
EINECS: 214-529-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR