Methyl-3-hydroxy-2-naphthoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-hydroxy-2-naphthoate

M-Clarity™ Qualitätsstufe = MQ100

167045-5G
SIAL167045-5GEA 136 EUR
SIAL167045-5G
Methyl-3-hydroxy-2-naphthoat, Sigma-Aldrich®
Methyl-3-hydroxy-2-naphthoat
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,21 g/mol
MDL: MFCD00004099
CAS-Nummer: 883-99-8
EINECS: 212-936-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR