tert-Butylcarbamat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Carbamidsäure-tert-butylester
M-Clarity™ Qualitätsstufe = MQ200
167398-5G
SIAL167398-5GEA
47.8
EUR
SIAL167398-5G
tert-Butylcarbamat, Sigma-Aldrich®
tert-Butylcarbamat
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol |
MDL:
MFCD00007962 CAS-Nummer: 4248-19-5 EINECS: 224-209-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...