Methyl-4-tert-butylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-(tert-butyl)benzoate

M-Clarity™ Qualitätsstufe = MQ100

167533-25G
SIAL167533-25GEA 154 EUR
SIAL167533-25G
Methyl-4-tert-butylbenzoat, Sigma-Aldrich®
Methyl-4-tert-butylbenzoat
Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
MDL: MFCD00008835
CAS-Nummer: 26537-19-9
EINECS: 247-768-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR