4-tert-Butylbenzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-tert-Butylbenzol-1-carbohydrazid, 4-tert-Butylbenzoesäurehydrazid, 4-tert-Butylbenzohydrazid

M-Clarity™ Qualitätsstufe = MQ200

168114-25G
SIAL168114-25GEA 77.9 EUR
SIAL168114-25G
4-tert-Butylbenzhydrazid, Sigma-Aldrich®
4-tert-Butylbenzhydrazid
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
MDL: MFCD00014763
CAS-Nummer: 43100-38-5
EINECS: 256-090-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR