2-(Methylthio)benzothiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Methylthio)benzo[d]thiazole

M-Clarity™ Qualitätsstufe = MQ100

168653-50G
SIAL168653-50GEA 168 EUR
SIAL168653-50G
2-(Methylthio)benzothiazol, Sigma-Aldrich®
2-(Methylthio)benzothiazole
Formel: C₈H₇NS₂
Molecular Weight: 181,2779 g/mol
MDL: MFCD00005784
CAS-Nummer: 615-22-5
EINECS: 210-417-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR