1-Methyl-1-phenylhydrazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

170267-5G
SIAL170267-5GEA 139 EUR
SIAL170267-5G
1-Methyl-1-phenylhydrazin, Sigma-Aldrich®
1-Methyl-1-phenylhydrazine
Formel: C₇H₁₀N₂
MDL: MFCD00007627
CAS-Nummer: 618-40-6
EINECS: 210-547-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR