Dimethylazelat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,9-Nonandisäuredimethylester, Azelainsäuredimethylester

M-Clarity™ Qualitätsstufe = MQ100

171026-500ML
SIAL171026-500MLEA 126 EUR
SIAL171026-500ML
Dimethylazelat, Sigma-Aldrich®
Dimethylazelat
Formel: C₁₁H₂₀O₄
Molecular Weight: 216,28 g/mol
MDL: MFCD00025898
CAS-Nummer: 1732-10-1
EINECS: 217-060-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR