1,1-Cyclohexandiessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1-Cyclohexanediaceticacid

M-Clarity™ Qualitätsstufe = MQ100

171344-5G
SIAL171344-5GEA 638 EUR
SIAL171344-5G
1,1-Cyclohexandiessigsäure, Sigma-Aldrich®
1,1-Cyclohexandiessigsäure
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
MDL: MFCD00001519
CAS-Nummer: 4355-11-7
EINECS: 224-427-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR