2-Pentanamin ≥97%, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 2-Amylamine, 1-Methylbutylamine, 2-Pentylamine

171417-5G
SIAL171417-5GEA 52.7 EUR
SIAL171417-5G
2-Pentanamin ≥97%, Sigma-Aldrich®
2-Aminopentane
Formel: C₅H₁₃N
Molecular Weight: 87,17 g/mol
Flammpunkt: 35 °C
MDL: MFCD00008098
CAS-Nummer: 63493-28-7
EINECS: 210-886-2
UN: 1106
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR