Dehydrated culture media, ITC broth base
Lieferant: Merck
17156-500G
SIAL17156-500GEA
179
EUR
SIAL17156-500G
Dehydrated culture media, ITC broth base
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
ITC broth base can be used for the selective enrichment, enumeration of Y. enterocolitica.
- Final pH: 6,9 ±0,2 (25 °C)
Standards: Meets the specification of ISO 10273:2003.