3-Phenylpropionitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hydrocinnamonitrile

M-Clarity™ Qualitätsstufe = MQ100

171573-10G
SIAL171573-10GEA 124 EUR
SIAL171573-10G
3-Phenylpropionitril, Sigma-Aldrich®
3-Phenylpropionitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00001961
CAS-Nummer: 645-59-0
EINECS: 211-447-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR