Tetraethylenglycoldimethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5,8,11,14-Pentaoxapentadecane

M-Clarity™ Qualitätsstufe = MQ200

172405-1KG 172405-250G
SIAL172405-1KGEA 168 EUR
SIAL172405-1KG SIAL172405-250G
Tetraethylenglycoldimethylether, Sigma-Aldrich®
Tetraethylenglycoldimethylether
Formel: C₁₀H₂₂O₅
MDL: MFCD00008505
CAS-Nummer: 143-24-8
EINECS: 205-594-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR