4-(Trifluormethyl)aniliniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminobenzotrifluorid-Hydrochlorid, α,α,α-Trifluor-p-toluidin-Hydrochlorid, 4-Aminobenzotrifluorid HCl, α,α,α-Trifluor-p-toluidin Hydrochlorid, 4-Aminobenzotrifluorid Hydrochlorid, α,α,α-Trifluor-p-toluidin HCl

172871-5G
SIAL172871-5GEA 53.6 EUR
SIAL172871-5G
4-(Trifluormethyl)aniliniumchlorid, Sigma-Aldrich®
4-(Trifluormethyl)aniliniumchlorid
Formel: C₇H₇ClF₃N
Molecular Weight: 197,59 g/mol
Schmelzpunkt: 200 °C
MDL: MFCD00007897
CAS-Nummer: 90774-69-9
EINECS: 292-679-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR