2-Hydroxyethylmethacrylat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Methylpropensäure(2-hydroxyethyl)ester, Methacrylsäure(2-hydroxyethyl)ester

17348-250ML-F
SIAL17348-250ML-FEA 133 EUR
SIAL17348-250ML-F
2-Hydroxyethylmethacrylat, Sigma-Aldrich®
2-Hydroxyethylmethacrylat
Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
MDL: MFCD00002863
CAS-Nummer: 868-77-9
EINECS: 212-782-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR