(±)-4-Brommandelsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(±)-2-(4-Bromphenyl)-2-hydroxyessigsäure, (±)-4-Bromphenylglykolsäure, (±)-2-(4-Bromphenyl)-2-hydroxyethansäure
17520-10G
SIAL17520-10GEA
168
EUR
SIAL17520-10G
(±)-4-Brommandelsäure, Sigma-Aldrich®
(±)-4-Brommandelsäure
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Siedepunkt: 344 °C (1013 hPa) Schmelzpunkt: 115…118 °C |
MDL:
MFCD00004232 CAS-Nummer: 6940-50-7 EINECS: 230-085-1 Merck Index: 13,01409 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...