6-Methoxy-1-indanon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
175250-5G
SIAL175250-5GEA
17.9
EUR
SIAL175250-5G
6-Methoxy-1-indanon, Sigma-Aldrich®
6-Methoxy-1-indanon
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Siedepunkt: 270 °C (1013 hPa) Schmelzpunkt: 108 °C |
MDL:
MFCD00021232 CAS-Nummer: 13623-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...