Essigsäureanhydrid-[D6] (99% D), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

175641-1G 175641-5G
SIAL175641-1GEA 83.6 EUR
SIAL175641-1G SIAL175641-5G
Essigsäureanhydrid-[D6] (99% D), Sigma-Aldrich®
Essigsäureanhydrid-[D6]
Formel: C₄D₆O₃
MDL: MFCD00044225
CAS-Nummer: 16649-49-3
EINECS: 240-697-0
UN: 1715
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR