Malonsäure-[D4], Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Malonic acid-[²H₄]

M-Clarity™ Qualitätsstufe = MQ200

175854-10G
SIAL175854-10GEA 88.2 EUR
SIAL175854-10G
Malonsäure-[D4], Sigma-Aldrich®
Malonsäure-[D4]
Formel: C₃D₄O₄
Molecular Weight: 108,03 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 813-56-9
EINECS: 212-385-4
Merck Index: 13,05732

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR