Triethyloxoniumtetrafluoroborat 1 mol/l in Dichlormethan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
176230-100ML
SIAL176230-100MLEA
144
EUR
SIAL176230-100ML
Triethyloxoniumtetrafluoroborat 1 mol/l in Dichlormethan, Sigma-Aldrich®
Triethyloxonium tetrafluoroborate
Formel:
C₆H₁₅BF₄O Molecular Weight: 189,99 g/mol |
MDL:
MFCD00044423 CAS-Nummer: 368-39-8 EINECS: 206-705-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...