Nitroniumtetrafluoroborat 0.5 mol/l in Sulfolan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Nitryl tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

176303-200ML
SIAL176303-200MLEA 375 EUR
SIAL176303-200ML
Nitroniumtetrafluoroborat 0.5 mol/l in Sulfolan, Sigma-Aldrich®
Nitroniumtetrafluoroborat
Formel: BF₄NO₂
Molecular Weight: 132,81 g/mol
MDL: MFCD00011432
CAS-Nummer: 13826-86-3
EINECS: 237-533-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR