3-Bromfuran, Sigma-Aldrich®
Lieferant: Merck
176524-10G
176524-2.5G
SIAL176524-10GEA
756
EUR
SIAL176524-10G
SIAL176524-2.5G
3-Bromfuran, Sigma-Aldrich®
3-Bromofuran
Formel:
C₄H₃BrO Siedepunkt: 103 °C (1013 hPa) Dichte: 1,6957 g/cm³ (0 °C) |
MDL:
MFCD00005347 CAS-Nummer: 22037-28-1 EINECS: 244-744-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...