3-Chinolincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Cyanoquinoline

M-Clarity™ Qualitätsstufe = MQ100

177156-5G
SIAL177156-5GEA 217 EUR
SIAL177156-5G
3-Chinolincarbonitril, Sigma-Aldrich®
3-Chinolincarbonitril
Formel: C₁₀H₆N₂
Molecular Weight: 154,17 g/mol
MDL: MFCD00006766
CAS-Nummer: 34846-64-5
EINECS: 252-248-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR