N,N,3,5-Tetramethylanilin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N,N-Dimethyl-3,5-xylidin, 3,5,N,N-Tetramethylanilin
M-Clarity™ Qualitätsstufe = MQ200
177342-25G
SIAL177342-25GEA
150
EUR
SIAL177342-25G
N,N,3,5-Tetramethylanilin, Sigma-Aldrich®
N,N,3,5-Tetramethylanilin
Formel:
C₁₀H₁₅N Molecular Weight: 149.24 g/mol Siedepunkt: 88 Dichte: 0.9275 |
MDL:
MFCD00008307 CAS-Nummer: 4913-13-7 EINECS: 225-539-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...