4-Acetylbenzoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Acetophenon-4-carbonsäure
M-Clarity™ Qualitätsstufe = MQ100
177458-10G
177458-1G
SIAL177458-10GEA
153
EUR
SIAL177458-10G
SIAL177458-1G
4-Acetylbenzoesäure, Sigma-Aldrich®
4-Acetylbenzoesäure
Formel:
C₉H₈O₃ Molecular Weight: 164.16 g/mol Siedepunkt: 340 |
MDL:
MFCD00002561 CAS-Nummer: 586-89-0 EINECS: 209-588-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...