2,2,2-Trifluorethyltoluol-4-sulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoroethyl p-toluenesulphonate, 2,2,2-Trifluoroethyl 4-methylbenzenesulphonate

M-Clarity™ Qualitätsstufe = MQ100

177822-50G
SIAL177822-50GEA 88.3 EUR
SIAL177822-50G
2,2,2-Trifluorethyltoluol-4-sulfonat, Sigma-Aldrich®
2,2,2-Trifluorethyltoluol-4-sulfonat
Formel: C₉H₉F₃O₃S
Molecular Weight: 254,23 g/mol
MDL: MFCD00000443
CAS-Nummer: 433-06-7
EINECS: 207-085-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR