Glutarimid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Piperidine-2,6-dione
178098-25G
178098-5G
SIAL178098-25GEA
345
EUR
SIAL178098-25G
SIAL178098-5G
Glutarimid, Sigma-Aldrich®
Glutarimid
Formel:
C₅H₇NO₂ Molecular Weight: 113.12 g/mol |
MDL:
MFCD00006670 CAS-Nummer: 1121-89-7 EINECS: 214-340-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...