Glutarimid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperidine-2,6-dione

178098-25G 178098-5G
SIAL178098-25GEA 345 EUR
SIAL178098-25G SIAL178098-5G
Glutarimid, Sigma-Aldrich®
Glutarimid
Formel: C₅H₇NO₂
Molecular Weight: 113.12 g/mol
MDL: MFCD00006670
CAS-Nummer: 1121-89-7
EINECS: 214-340-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR