2,4,4-Trimethyl-2-oxazolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,4-Trimethyl-4,5-dihydrooxazole

178748-25G
SIAL178748-25GEA 48 EUR
SIAL178748-25G
2,4,4-Trimethyl-2-oxazolin, Sigma-Aldrich®
2,4,4-Trimethyl-2-oxazolin
Formel: C₆H₁₁NO
Molecular Weight: 113,16 g/mol
MDL: MFCD00005301
CAS-Nummer: 1772-43-6
EINECS: 217-201-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR