Dimethyl methylsuccinate ≥98%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4-dimethyl (2S)-2-methylbutanedioate, Dimethyl (2S)-2-methylsuccinate, (S)-(-)-Methylsuccinic acid dimethyl ester

M-Clarity™ Qualitätsstufe = MQ100

178799-10G
SIAL178799-10GEA 55.5 EUR
SIAL178799-10G
Dimethyl methylsuccinate ≥98%, Sigma-Aldrich®
Dimethyl methylsuccinate

Dimethyl methylsuccinate is used in the synthesis of β-half esters.

Formel: C₇H₁₂O₄
Molecular Weight: 160,168 g/mol
Siedepunkt: 196 °C (Lit.)
Dichte: 1,076 g/ml (25 °C) (Lit.)
Flammpunkt: 83 °C
MDL: MFCD00008449
CAS-Nummer: 1604-11-1
EINECS: 216-506-1

Bestellen

Spezifikation Testergebnisse

Assay 98%
Refractive index n20/D 1.42 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR