(±)-β-Methylphenethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Phenylpropylamine

180076-5G
SIAL180076-5GEA 120 EUR
SIAL180076-5G
(±)-β-Methylphenethylamin, Sigma-Aldrich®
(±)-β-Methylphenethylamin
Formel: C₉H₁₃N
MDL: MFCD00008144
CAS-Nummer: 582-22-9
EINECS: 209-479-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR