5,6,7,8-Tetrahydro-2-naphthalinol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

180459-10G
SIAL180459-10GEA 73.7 EUR
SIAL180459-10G
5,6,7,8-Tetrahydro-2-naphthalinol, Sigma-Aldrich®
5,6,7,8-Tetrahydro-2-naphthol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
MDL: MFCD00001738
CAS-Nummer: 1125-78-6
EINECS: 214-410-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR