(2E,4E)-2,4-Decadienal, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

180513-5G
SIAL180513-5GEA 129 EUR
SIAL180513-5G
(2E,4E)-2,4-Decadienal, Sigma-Aldrich®
trans,trans-2,4-Decadienal
Formel: C₁₀H₁₆O
MDL: MFCD00007007
CAS-Nummer: 25152-84-5
EINECS: 246-668-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR