(2-Methoxyphenyl)acetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methoxybenzylcyanid

M-Clarity™ Qualitätsstufe = MQ100

180645-50G
SIAL180645-50GEA 46 EUR
SIAL180645-50G
(2-Methoxyphenyl)acetonitril, Sigma-Aldrich®
(2-Methoxyphenyl)acetonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00001902
CAS-Nummer: 7035-03-2
EINECS: 230-314-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR