(3-Brompropyl)trimethoxysilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide

18265-5ML
SIAL18265-5MLEA 212 EUR
SIAL18265-5ML
(3-Brompropyl)trimethoxysilan, Sigma-Aldrich®
(3-Brompropyl)trimethoxysilan
Formel: BrH₂CCH₂CH₂Si(OCH₃)₃
Molecular Weight: 243,17 g/mol
MDL: MFCD00053894
CAS-Nummer: 51826-90-5
EINECS: 257-453-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR