trans,trans-2,4-Hexadien-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans,trans-Hexa-2,4-dien-1-ol, trans,trans-1-Hydroxy-2,4-hexadien

MQ100

183059-10G 183059-50G
SIAL183059-10GEA 76 EUR
SIAL183059-10G SIAL183059-50G
trans,trans-2,4-Hexadien-1-ol, Sigma-Aldrich®
trans,trans-2,4-Hexadien-1-ol
Formel: C₆H₁₀O
Molecular Weight: 98.14 g/mol
MDL: MFCD00002925
CAS-Nummer: 17102-64-6
EINECS: 241-173-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR