1-Methylisatin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Methylindolin-2,3-dion, 1-Methyl-1H-indol-2,3-dion, N-Methyl-2,3-dioxoindolin, N-Methyl-2,3-dihydroindol-2,3-dion, N-Methyl-1H-indol-2,3-dion, N-Methylindolin-2,3-dion, 1-Methyl-2,3-indolindion, N-Methylisatin, 1-Methyl-2,3-dioxoindolin, N-Methyl-2,3-indolindion, 1-Methyl-2,3-dihydroindol-2,3-dion
183075-10G
183075-1G
SIAL183075-10GEA
59.6
EUR
SIAL183075-10G
SIAL183075-1G
1-Methylisatin, Sigma-Aldrich®
1-Methylisatin
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Schmelzpunkt: 139 Dichte: 1.4 g/cm³ (20 °C) |
MDL:
MFCD00005812 CAS-Nummer: 2058-74-4 EINECS: 218-164-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...