3',4'-Dichlorpropiophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(3,4-Dichlorphenyl)-1-propanon
183415-5G
SIAL183415-5GEA
467
EUR
SIAL183415-5G
3',4'-Dichlorpropiophenon, Sigma-Aldrich®
3',4'-Dichlorpropiophenon
Formel:
C₉H₈Cl₂O Molecular Weight: 203,07 g/mol Siedepunkt: 257 °C (1013 hPa) Schmelzpunkt: 44 °C |
MDL:
MFCD00000554 CAS-Nummer: 6582-42-9 EINECS: 229-511-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...