2-(1-Naphthalin)ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Naphthalen-1-yl)ethanol, 2-(1-Naphthyl)ethanol

M-Clarity™ Qualitätsstufe = MQ100

183458-10G
SIAL183458-10GEA 73.9 EUR
SIAL183458-10G
2-(1-Naphthalin)ethanol, Sigma-Aldrich®
2-(1-Naphthalin)ethanol
Formel: C₁₂H₁₂O
Molecular Weight: 172,23 g/mol
MDL: MFCD00004050
CAS-Nummer: 773-99-9
EINECS: 212-260-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR