N-Phenyl-m-toluidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methyldiphenylamine

M-Clarity™ Qualitätsstufe = MQ100

183512-10G
SIAL183512-10GEA 44.4 EUR
SIAL183512-10G
N-Phenyl-m-toluidin, Sigma-Aldrich®
N-Phenyl-m-toluidin
Formel: C₁₃H₁₃N
Molecular Weight: 183,25 g/mol
MDL: MFCD00008530
CAS-Nummer: 1205-64-7
EINECS: 214-885-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR