7-(Trifluormethyl)-4-chinolinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-7-(trifluoromethyl)quinoline

184217-10G
SIAL184217-10GEA 149 EUR
SIAL184217-10G
7-(Trifluormethyl)-4-chinolinol, Sigma-Aldrich®
7-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,1560 g/mol
MDL: MFCD00006779
CAS-Nummer: 322-97-4
EINECS: 206-298-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR