(4-Fluorphenyl)methylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Fluor-4-(methylsulfonyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

184330-5G
SIAL184330-5GEA 38.7 EUR
SIAL184330-5G
(4-Fluorphenyl)methylsulfon, Sigma-Aldrich®
(4-Fluorphenyl)methylsulfon
Formel: C₇H₇FO₂S
Molecular Weight: 174,2 g/mol
MDL: MFCD00039753
CAS-Nummer: 455-15-2
EINECS: 207-237-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR